Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334962.1 | NP_001323118.1 | 3 - 4 | 951 | 64 | |
NM_001334963.1 | NP_001323117.1 | 4 - 5 | 615 | 64 | ||
NM_001334964.1 | NP_001323120.1 | 3 - 4 | 705 | 64 | ||
NM_001334965.1 | NP_001323116.1 | 3 - 4 | 691 | 64 | ||
NM_001334966.1 | NP_001323119.1 | 4 - 5 | 583 | 64 | ||
NM_106668.6 | NP_178136.1 | 4 - 5 | 598 | 64 | ||
GenBank mRNA | AK175552.1 | - | 4 - 5 | 548 | 64 | |
BT010484.1 | - | 3 - 4 | 256 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |