Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334999.1 | NP_001319430.1 | 10 - 11 | 3080 | 84 | |
NM_106714.2 | NP_178181.1 | 11 - 12 | 3012 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |