Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPTOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111719.3 | NP_566335.1 | 2 - 3 | 902 | 137 | |
GenBank mRNA | AY059838.1 | - | 2 - 3 | 802 | 137 | |
AY769948.1 | - | 2 - 3 | 802 | 137 | ||
BT008813.1 | - | 2 - 3 | 413 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |