Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125130.1 | NP_001118602.1 | 11 - 12 | 2440 | 77 | |
NM_001202915.1 | NP_001189844.1 | 11 - 12 | 2321 | 77 | ||
NM_111744.5 | NP_566343.1 | 11 - 12 | 2566 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |