Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337807.1 | NP_001326525.1 | 3 - 4 | 1170 | 70 | |
NM_111756.3 | NP_187533.1 | 4 - 5 | 795 | 70 | ||
GenBank mRNA | AK117639.1 | - | 4 - 5 | 682 | 70 | |
BT004971.1 | - | 4 - 5 | 617 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |