Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111764.4 | NP_187541.1 | 7 - 8 | 1031 | 102 | |
GenBank mRNA | AY062703.1 | - | 7 - 8 | 996 | 102 | |
AY093373.1 | - | 7 - 8 | 810 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |