Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB3R-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111771.4 | NP_566350.1 | 1 - 2 | 223 | 101 | |
GenBank mRNA | AY142649.1 | - | 1 - 2 | 42 | 101 | |
AY519651.1 | - | 1 - 2 | 42 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |