Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337815.1 | NP_001327181.1 | 2 - 3 | 298 | 68 | |
NM_111773.4 | NP_187550.1 | 1 - 2 | 451 | 68 | ||
GenBank mRNA | AY037263.1 | - | 1 - 2 | 94 | 68 | |
AY077669.1 | - | 1 - 2 | 65 | 68 | ||
U15108.1 | - | 1 - 2 | 68 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |