Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035587.3 | NP_001030664.1 | 5 - 6 | 476 | 82 | |
NM_111798.4 | NP_187575.2 | 4 - 5 | 559 | 82 | ||
GenBank mRNA | AK176821.1 | - | 5 - 6 | 476 | 82 | |
AK176908.1 | - | 4 - 5 | 559 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |