Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABC2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202921.2 | NP_001189850.1 | 4 - 5 | 348 | 120 | |
NM_001202922.1 | NP_001189851.1 | 4 - 5 | 341 | 120 | ||
NM_001337848.1 | NP_001326433.1 | 3 - 4 | 596 | 120 | ||
NM_001337849.1 | NP_001326432.1 | 4 - 5 | 353 | 120 | ||
NM_111826.4 | NP_187602.1 | 3 - 4 | 647 | 120 | ||
GenBank mRNA | BT012209.1 | - | 3 - 4 | 365 | 120 | |
BT012396.1 | - | 3 - 4 | 266 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |