Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337853.1 | NP_001326647.1 | 9 - 10 | 2834 | 89 | |
NM_111836.5 | NP_187612.5 | 8 - 9 | 2704 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |