Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337859.1 | NP_001326965.1 | 3 - 4 | 997 | 93 | |
NM_111850.4 | NP_187626.1 | 3 - 4 | 904 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |