Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111876.5 | NP_187652.1 | 5 - 6 | 1127 | 78 | |
GenBank mRNA | AY091767.1 | - | 5 - 6 | 944 | 78 | |
AY149954.1 | - | 5 - 6 | 923 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |