Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111903.3 | NP_187678.1 | 3 - 4 | 778 | 120 | |
GenBank mRNA | AY035125.1 | - | 3 - 4 | 719 | 120 | |
AY113917.1 | - | 3 - 4 | 636 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |