Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337892.1 | NP_001326565.1 | 14 - 15 | 2297 | 68 | |
NM_001337893.1 | NP_001326566.1 | 14 - 15 | 2446 | 68 | ||
NM_001337894.1 | NP_001326567.1 | 13 - 14 | 1571 | 68 | ||
NM_111911.5 | NP_187685.1 | 14 - 15 | 2109 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |