Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035593.2 | NP_001030670.1 | 2 - 3 | 407 | 93 | |
NM_111929.4 | NP_187703.1 | 2 - 3 | 421 | 93 | ||
GenBank mRNA | AY059807.1 | - | 2 - 3 | 373 | 93 | |
AY072495.1 | - | 2 - 3 | 346 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |