Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111942.4 | NP_187716.1 | 6 - 7 | 828 | 120 | |
GenBank mRNA | AJ312192.1 | - | 6 - 7 | 687 | 120 | |
AK175807.1 | - | 6 - 7 | 708 | 120 | ||
AK175899.1 | - | 6 - 7 | 708 | 120 | ||
AK175901.1 | - | 6 - 7 | 708 | 120 | ||
AK175910.1 | - | 6 - 7 | 708 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |