Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202930.1 | NP_001189859.1 | 8 - 9 | 945 | 108 | |
NM_111962.5 | NP_187736.1 | 8 - 9 | 933 | 108 | ||
GenBank mRNA | AK228032.1 | - | 8 - 9 | 865 | 108 | |
AY258412.1 | - | 8 - 9 | 766 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |