Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111981.5 | NP_187755.2 | 2 - 3 | 262 | 75 | |
GenBank mRNA | AK221911.1 | - | 2 - 3 | 208 | 75 | |
AY224595.1 | - | 2 - 3 | 139 | 75 | ||
BT022049.1 | - | 2 - 3 | 203 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |