Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 12 - 13 | 2201 | 119 | |
NM_202557.2 | NP_974286.1 | 7 - 8 | 1380 | 119 | ||
GenBank mRNA | AK221314.1 | - | 12 - 13 | 1927 | 119 | |
KJ138828.1 | - | 11 - 12 | 1566 | 119 | ||
U62135.1 | - | 12 - 13 | 1916 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |