Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111987.4 | NP_187761.1 | 15 - 16 | 2633 | 67 | |
NM_202557.2 | NP_974286.1 | 10 - 11 | 1812 | 67 | ||
GenBank mRNA | AK221314.1 | - | 15 - 16 | 2359 | 67 | |
KJ138828.1 | - | 14 - 15 | 1998 | 67 | ||
U62135.1 | - | 15 - 16 | 2348 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |