Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338905.1 | NP_001326065.1 | 13 - 14 | 1561 | 92 | |
NM_001338906.1 | NP_001326062.1 | 13 - 14 | 1471 | 92 | ||
NM_001338907.1 | NP_001326064.1 | 13 - 14 | 1408 | 92 | ||
NM_001338908.1 | NP_001326061.1 | 12 - 13 | 1434 | 92 | ||
NM_001338909.1 | NP_001326060.1 | 13 - 14 | 1562 | 92 | ||
NM_001338910.1 | NP_001326063.1 | 13 - 14 | 1423 | 92 | ||
NM_113689.2 | NP_189410.2 | 12 - 13 | 1434 | 92 | ||
GenBank mRNA | AY822649.1 | - | 12 - 13 | 1285 | 92 | |
AY960701.1 | - | 12 - 13 | 1434 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |