Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338905.1 | NP_001326065.1 | 17 - 18 | 1889 | 115 | |
NM_001338906.1 | NP_001326062.1 | 17 - 18 | 1799 | 115 | ||
NM_001338907.1 | NP_001326064.1 | 17 - 18 | 1736 | 115 | ||
NM_001338908.1 | NP_001326061.1 | 16 - 17 | 1762 | 115 | ||
NM_001338909.1 | NP_001326060.1 | 17 - 18 | 1890 | 115 | ||
NM_001338910.1 | NP_001326063.1 | 17 - 18 | 1751 | 115 | ||
NM_113689.2 | NP_189410.2 | 16 - 17 | 1762 | 115 | ||
GenBank mRNA | AY822649.1 | - | 16 - 17 | 1613 | 115 | |
AY960701.1 | - | 16 - 17 | 1762 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |