Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338905.1 | NP_001326065.1 | 20 - 21 | 2396 | 60 | |
NM_001338906.1 | NP_001326062.1 | 20 - 21 | 2306 | 60 | ||
NM_001338907.1 | NP_001326064.1 | 20 - 21 | 2243 | 60 | ||
NM_001338908.1 | NP_001326061.1 | 19 - 20 | 2269 | 60 | ||
NM_001338909.1 | NP_001326060.1 | 20 - 21 | 2397 | 60 | ||
NM_001338910.1 | NP_001326063.1 | 20 - 21 | 2258 | 60 | ||
NM_113689.2 | NP_189410.2 | 19 - 20 | 2269 | 60 | ||
GenBank mRNA | AY822649.1 | - | 19 - 20 | 2120 | 60 | |
AY960701.1 | - | 19 - 20 | 2269 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |