Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338905.1 | NP_001326065.1 | 3 - 4 | 523 | 111 | |
NM_001338906.1 | NP_001326062.1 | 3 - 4 | 433 | 111 | ||
NM_001338907.1 | NP_001326064.1 | 3 - 4 | 370 | 111 | ||
NM_001338908.1 | NP_001326061.1 | 2 - 3 | 396 | 111 | ||
NM_001338909.1 | NP_001326060.1 | 3 - 4 | 524 | 111 | ||
NM_001338910.1 | NP_001326063.1 | 3 - 4 | 385 | 111 | ||
NM_113689.2 | NP_189410.2 | 2 - 3 | 396 | 111 | ||
GenBank mRNA | AY822649.1 | - | 2 - 3 | 247 | 111 | |
AY960701.1 | - | 2 - 3 | 396 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |