Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338949.1 | NP_001327074.1 | 1 - 2 | 193 | 112 | |
NM_113740.6 | NP_189461.1 | 1 - 2 | 252 | 112 | ||
GenBank mRNA | AF139098.1 | - | 1 - 2 | 166 | 112 | |
AK176745.1 | - | 1 - 2 | 187 | 112 | ||
BT024832.1 | - | 1 - 2 | 110 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |