Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113794.4 | NP_189515.1 | 9 - 10 | 1017 | 114 | |
GenBank mRNA | BT002507.1 | - | 9 - 10 | 1011 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |