Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81D11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113795.2 | NP_189516.1 | 2 - 3 | 917 | 91 | |
GenBank mRNA | AY050849.1 | - | 2 - 3 | 917 | 91 | |
AY113869.1 | - | 2 - 3 | 916 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |