Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338993.1 | NP_001326021.1 | 1 - 2 | 199 | 97 | |
NM_113818.4 | NP_189539.1 | 1 - 2 | 292 | 97 | ||
GenBank mRNA | AK118926.1 | - | 1 - 2 | 197 | 97 | |
BT005690.1 | - | 1 - 2 | 58 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |