Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113824.3 | NP_189545.1 | 2 - 3 | 843 | 97 | |
GenBank mRNA | BT025711.1 | - | 2 - 3 | 460 | 97 | |
U30478.1 | - | 2 - 3 | 560 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |