Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113910.5 | NP_566851.1 | 4 - 5 | 961 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |