Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAXT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339214.1 | NP_001327486.1 | 1 - 2 | 473 | 67 | |
NM_114434.2 | NP_190151.1 | 1 - 2 | 473 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |