Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 17 - 18 | 2795 | 75 | |
NM_114447.4 | NP_190164.1 | 18 - 19 | 2793 | 75 | ||
GenBank mRNA | AF030864.1 | - | 18 - 19 | 2648 | 75 | |
AF360218.1 | - | 17 - 18 | 2602 | 75 | ||
AY040062.1 | - | 17 - 18 | 2585 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |