Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114458.4 | NP_190175.2 | 4 - 5 | 1356 | 68 | |
GenBank mRNA | AK229694.1 | - | 4 - 5 | 1253 | 68 | |
BT008583.1 | - | 4 - 5 | 1237 | 68 | ||
BT008683.1 | - | 4 - 5 | 1253 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |