Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114479.5 | NP_190196.1 | 3 - 4 | 539 | 75 | |
GenBank mRNA | AF020715.1 | - | 3 - 4 | 475 | 75 | |
AY056233.1 | - | 3 - 4 | 480 | 75 | ||
AY117252.1 | - | 3 - 4 | 470 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |