Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114487.4 | NP_190204.2 | 8 - 9 | 936 | 77 | |
GenBank mRNA | BT011224.1 | - | 8 - 9 | 877 | 77 | |
BT012152.1 | - | 8 - 9 | 851 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |