Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339237.1 | NP_001327061.1 | 3 - 4 | 1237 | 74 | |
NM_001339238.1 | NP_001327062.1 | 4 - 5 | 1159 | 74 | ||
NM_001339239.1 | NP_001327064.1 | 4 - 5 | 1159 | 74 | ||
NM_001339240.1 | NP_001327065.1 | 3 - 4 | 415 | 74 | ||
NM_001339241.1 | NP_001327063.1 | 3 - 4 | 394 | 74 | ||
NM_114489.5 | NP_190206.2 | 4 - 5 | 1159 | 74 | ||
GenBank mRNA | AY070078.1 | - | 4 - 5 | 316 | 74 | |
AY117353.1 | - | 4 - 5 | 299 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |