Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035744.2 | NP_001030821.1 | 4 - 5 | 1482 | 70 | |
NM_001035745.2 | NP_001030822.2 | 4 - 5 | 1149 | 70 | ||
NM_001339266.1 | NP_001326037.1 | 4 - 5 | 1259 | 70 | ||
NM_001339267.1 | NP_001326038.1 | 2 - 3 | 780 | 70 | ||
NM_114526.3 | NP_190243.2 | 4 - 5 | 1164 | 70 | ||
GenBank mRNA | AJ630475.1 | - | 3 - 4 | 788 | 70 | |
AK229705.1 | - | 4 - 5 | 1071 | 70 | ||
AY395985.1 | - | 4 - 5 | 1120 | 70 | ||
AY519541.1 | - | 3 - 4 | 788 | 70 | ||
AY568647.1 | - | 3 - 4 | 788 | 70 | ||
BT033142.1 | - | 3 - 4 | 791 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |