Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT76E12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114533.2 | NP_566885.1 | 1 - 2 | 795 | 86 | |
GenBank mRNA | AY048297.1 | - | 1 - 2 | 633 | 86 | |
AY120731.1 | - | 1 - 2 | 532 | 86 | ||
BT000356.1 | - | 1 - 2 | 490 | 86 | ||
BT002638.1 | - | 1 - 2 | 490 | 86 | ||
KJ138803.1 | - | 1 - 2 | 490 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |