Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC75-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114541.3 | NP_190258.1 | 6 - 7 | 2330 | 63 | |
GenBank mRNA | AY127014.1 | - | 6 - 7 | 2262 | 63 | |
BT006358.1 | - | 6 - 7 | 2215 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |