Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114564.3 | NP_190281.1 | 8 - 9 | 1531 | 62 | |
GenBank mRNA | AY090236.1 | - | 8 - 9 | 1252 | 62 | |
BT003012.1 | - | 7 - 8 | 1120 | 62 | ||
KJ138797.1 | - | 7 - 8 | 1120 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |