Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114573.3 | NP_566889.1 | 8 - 9 | 1672 | 72 | |
GenBank mRNA | AY099737.1 | - | 8 - 9 | 1613 | 72 | |
BT000368.1 | - | 8 - 9 | 1599 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |