Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161195.1 | NP_001154667.1 | 4 - 5 | 622 | 112 | |
NM_114603.3 | NP_190319.1 | 4 - 5 | 622 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |