Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339310.1 | NP_001325474.1 | 8 - 9 | 2177 | 146 | |
NM_114614.4 | NP_190330.1 | 8 - 9 | 2114 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |