Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114618.4 | NP_190334.1 | 1 - 2 | 302 | 109 | |
GenBank mRNA | AF079503.1 | - | 1 - 2 | 239 | 109 | |
AY065152.1 | - | 1 - 2 | 227 | 109 | ||
AY128757.1 | - | 1 - 2 | 142 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |