Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114642.4 | NP_190358.3 | 2 - 3 | 568 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |