Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339335.1 | NP_001327229.1 | 11 - 12 | 1859 | 118 | |
NM_001339336.1 | NP_001327230.1 | 11 - 12 | 1859 | 118 | ||
NM_114645.3 | NP_190361.2 | 10 - 11 | 1698 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |