Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114645.3 | NP_190361.2 | 3 - 4 | 519 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |