Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339337.1 | NP_001327142.1 | 6 - 7 | 1146 | 110 | |
NM_001339338.1 | NP_001327141.1 | 7 - 8 | 1055 | 110 | ||
NM_114647.4 | NP_190363.3 | 7 - 8 | 1055 | 110 | ||
GenBank mRNA | AY099665.1 | - | 7 - 8 | 1054 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |