Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114732.5 | NP_190442.1 | 2 - 3 | 475 | 126 | |
GenBank mRNA | AK229328.1 | - | 2 - 3 | 423 | 126 | |
BT025324.1 | - | 2 - 3 | 349 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |